Ethyl 2-(2-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

93378-5G 93378-25G 93378-1G 93378-500MG
77630-884EA 1441 USD
77630-884 77636-252 77640-204 77652-214
Ethyl 2-(2-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-(methoxymethoxy)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 238.237 g/mol
MW: C₁₂H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD34818533
CAS Number: 1821472-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR