4-(5-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

93354-5G 93354-25G 93354-1G 93354-500MG
77630-882EA 1441 USD
77630-882 77636-250 77640-200 77652-210
4-(5-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(5-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 240.271 g/mol
MW: C₁₃H₁₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD34773508
CAS Number: 2918830-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR