4-(2,5-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

93280-5G 93280-25G 93280-1G 93280-500MG
77633-852EA 1206 USD
77633-852 77660-554 77644-208 77648-198
4-(2,5-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2,5-Difluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18296196
CAS Number: 1344216-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR