1-(4-Chloro-2-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93175-5G 93175-500MG 93175-25G 93175-1G
77633-850EA 1357 USD
77633-850 77653-126 77657-070 77644-206
1-(4-Chloro-2-methylphenyl)cyclobutanol ≥95%
1-(4-Chloro-2-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD34773446
CAS Number: 2160711-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR