1-(2,3-Dimethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93153-5G 93153-1G 93153-25G 93153-500MG
77630-874EA 1441 USD
77630-874 77640-198 77660-550 77652-208
1-(2,3-Dimethylphenyl)cyclobutanol ≥95%
1-(2,3-Dimethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 176.255 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD21788747
CAS Number: 1526494-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR