1-(4-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

93151-5G 93151-1G 93151-25G 93151-500MG
77630-872EA 1441 USD
77630-872 77640-196 77657-068 77648-190
1-(4-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%
1-(4-Fluoro-2-methoxyphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 210.245 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34773431
CAS Number: 2918946-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR