(2-Methyl-1,4-phenylene)bis(methylsulfane) ≥95%

Supplier: AOBChem USA

Warning

92929-5G 92929-1G 92929-25G 92929-500MG
77633-848EA 1441 USD
77633-848 77640-194 77657-064 77652-204
(2-Methyl-1,4-phenylene)bis(methylsulfane) ≥95%
(2-Methyl-1,4-phenylene)bis(methylsulfane)

Precaución: For Research Use Only

Formula: C₉H₁₂S₂
MW: 184.322 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25967763
CAS Number: 84910-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR