2-(3,4-Difluorophenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

92584-5G 92584-1G 92584-25G 92584-500MG
77633-846EA 712 USD
77633-846 77640-192 77657-062 77652-202
2-(3,4-Difluorophenyl)-2-oxoacetic acid ≥97%
2-(3,4-Difluorophenyl)-2-oxoacetic acid

Precaución: For Research Use Only

Formula: C₈H₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD07698680
CAS Number: 890097-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR