2-(2-Methoxyphenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

92499-5G 92499-1G 92499-25G 92499-500MG
77633-844EA 698 USD
77633-844 77640-190 77660-546 77648-186
2-(2-Methoxyphenyl)-2-oxoacetic acid ≥97%
2-(2-Methoxyphenyl)-2-oxoacetic acid

Precaución: For Research Use Only

Formula: 180.157 g/mol
MW: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD02666071
CAS Number: 26767-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR