Mesitylglyoxylic acid ≥95%

Supplier: AOBChem USA

Warning

92495-5G 92495-25G
77635-368EA 59 USD
77635-368 77660-544
Mesitylglyoxylic acid ≥95%
Mesitylglyoxylic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00151834
CAS Number: 3112-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR