2-(2-Chlorophenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

92492-5G 92492-25G 92492-1G 92492-500MG
77635-366EA 314 USD
77635-366 77657-060 77644-198 77652-200
2-(2-Chlorophenyl)-2-oxoacetic acid ≥97%
2-(2-Chlorophenyl)-2-oxoacetic acid

Precaución: For Research Use Only

Formula: C₈H₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD06410833
CAS Number: 26118-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR