N,N-Diethyl-6-methylnicotinamide ≥95%

Supplier: AOBChem USA

Warning

92291-5G 92291-1G 92291-25G 92291-500MG
77633-842EA 1207 USD
77633-842 77640-188 77657-058 77648-184
N,N-Diethyl-6-methylnicotinamide ≥95%
N,N-Diethyl-6-methylnicotinamide

Precaución: For Research Use Only

Formula: 192.258 g/mol
MW: C₁₁H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD11800829
CAS Number: 50385-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR