1-(2-Bromo-6-chlorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

92090-5G 92090-25G 92090-1G 92090-500MG
77630-866EA 1441 USD
77630-866 77660-538 77648-178 77648-180
1-(2-Bromo-6-chlorophenyl)ethanol ≥97%
1-(2-Bromo-6-chlorophenyl)ethanol

Precaución: For Research Use Only

Formula: 235.506 g/mol
MW: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD28061442
CAS Number: 1175300-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR