2-Ethyl-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

92058-1G 92058-5G 92058-500MG
77635-362EA 867 USD
77635-362 77657-054 77648-176
2-Ethyl-3-fluorobenzaldehyde ≥97%
2-Ethyl-3-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD28969420
CAS Number: 1289176-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR