2-Ethyl-3-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

92055-1G 92055-500MG 92055-5G
77635-360EA 837 USD
77635-360 77653-124 77657-052
2-Ethyl-3-fluorobenzoic acid ≥95%
2-Ethyl-3-fluorobenzoic acid

Precaución: For Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD18411930
CAS Number: 1261884-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR