3-Hydroxy-2,6-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

91649-1G 91649-5G 91649-500MG
77635-358EA 811 USD
77635-358 77660-536 77648-174
3-Hydroxy-2,6-dimethylbenzaldehyde ≥95%
3-Hydroxy-2,6-dimethylbenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD16249771
CAS Number: 179554-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR