6,7-Dichloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

91585-5G 91585-1G 91585-25G 91585-500MG
77633-840EA 1158 USD
77633-840 77640-184 77657-050 77648-172
6,7-Dichloro-1H-indazole ≥95%
6,7-Dichloro-1H-indazole

Precaución: For Research Use Only

Formula: 187.026 g/mol
MW: C₇H₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD11878112
CAS Number: 72521-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR