Ethyl 2-(benzo[b]thiophen-2-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

91385-5G 91385-25G 91385-1G 91385-500MG
77630-864EA 1441 USD
77630-864 77636-248 77640-182 77652-192
Ethyl 2-(benzo[b]thiophen-2-yl)-2-oxoacetate ≥95%
Ethyl 2-(benzo[b]thiophen-2-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 234.271 g/mol
MW: C₁₂H₁₀O₃S
Storage Temperature: Ambient
MDL Number: MFCD23133885
CAS Number: 98800-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR