Tert-butyl benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

91384-5G 91384-25G 91384-500MG 91384-1G
77630-862EA 1441 USD
77630-862 77636-246 77653-122 77644-192
Tert-butyl benzo[b]thiophene-2-carboxylate ≥95%
Tert-butyl benzo[b]thiophene-2-carboxylate

Precaución: For Research Use Only

Formula: 234.314 g/mol
MW: C₁₃H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD19388693
CAS Number: 1406637-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR