Ethyl-5-chlorothiophene-2-glyoxylate ≥95%

Supplier: AOBChem USA

Warning

91380-5G 91380-25G 91380-500MG 91380-1G
77633-838EA 1180 USD
77633-838 77657-048 77648-168 77648-166
Ethyl-5-chlorothiophene-2-glyoxylate ≥95%
Ethyl-5-chlorothiophene-2-glyoxylate

Precaución: For Research Use Only

Formula: C₈H₇ClO₃S
Storage Temperature: Ambient
MDL Number: MFCD07782946
CAS Number: 34036-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR