(2-Chloro-4-(methylsulfonyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

91167-1G 91167-5G 91167-500MG
77635-356EA 821 USD
77635-356 77660-530 77652-188
(2-Chloro-4-(methylsulfonyl)phenyl)boronic acid ≥95%
(2-Chloro-4-(methylsulfonyl)phenyl)boronic acid

Precaución: For Research Use Only

Formula: 234.465 g/mol
MW: C₇H₈BClO₄S
Storage Temperature: Ambient
MDL Number: MFCD23380966
CAS Number: 1262662-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR