Tert-butyl 2-methyl-4-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

91099-5G 91099-1G 91099-500MG
77660-528EA 2288 USD
77660-528 77648-162 77648-164
Tert-butyl 2-methyl-4-(methylthio)benzoate ≥95%
Tert-butyl 2-methyl-4-(methylthio)benzoate

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD34772623
CAS Number: 2918931-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR