2,6-Dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

91077-5G 91077-25G
77635-354EA 128 USD
77635-354 77660-526
2,6-Dimethylbenzamide ≥95%
2,6-Dimethylbenzamide

Precaución: For Research Use Only

Formula: C₉H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00052921
CAS Number: 55321-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR