2-Bromo-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

91038-1G 91038-5G 91038-500MG
77640-178EA 519 USD
77640-178 77660-524 77648-160
2-Bromo-6-(trifluoromethyl)benzamide ≥97%
2-Bromo-6-(trifluoromethyl)benzamide

Precaución: For Research Use Only

Formula: C₈H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD06253762
CAS Number: 610260-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR