(7-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
90701-1G
90701-5G
90701-500MG
77640-176EA
627
USD
77640-176
77660-522
77648-158
(7-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
(7-Methoxybenzo[d][1,3]dioxol-5-yl)boronic acid
Precaución: For Research Use Only
Formula:
195.965 g/mol MW: C₈H₉BO₅ Storage Temperature: Ambient |
MDL Number:
MFCD34772431 CAS Number: 221243-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...