5-Bromo-2-chloro-1,3-diisopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

90603-5G 90603-25G 90603-1G
77635-352EA 188 USD
77635-352 77660-518 77644-188
5-Bromo-2-chloro-1,3-diisopropylbenzene ≥97%
5-Bromo-2-chloro-1,3-diisopropylbenzene

Precaución: For Research Use Only

Formula: 275.612 g/mol
MW: C₁₂H₁₆BrCl
Storage Temperature: Ambient
MDL Number: MFCD34772424
CAS Number: 2918876-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR