2-Chloro-1,3-diisopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

90601-5G 90601-25G 90601-1G 90601-500MG
77633-832EA 1115 USD
77633-832 77660-516 77644-186 77648-156
2-Chloro-1,3-diisopropylbenzene ≥95%
2-Chloro-1,3-diisopropylbenzene

Precaución: For Research Use Only

Formula: 196.716 g/mol
MW: C₁₂H₁₇Cl
Storage Temperature: Ambient
MDL Number: MFCD18971516
CAS Number: 54845-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR