5-(Benzyloxy)picolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

90124-5G 90124-1G 90124-500MG
77630-860EA 1797 USD
77630-860 77644-182 77652-182
5-(Benzyloxy)picolinaldehyde ≥97%
5-(Benzyloxy)picolinaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD10697537
CAS Number: 59781-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR