1-(2,3-Dichloropyridin-4-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

90069-5G 90069-1G 90069-25G 90069-500MG
77630-858EA 1441 USD
77630-858 77640-174 77660-510 77648-154
1-(2,3-Dichloropyridin-4-yl)cyclobutanol ≥95%
1-(2,3-Dichloropyridin-4-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₉H₉Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD34772127
CAS Number: 2918850-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR