2-Bromo-1-(cyclopropylmethoxy)-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

89973-5G 89973-25G 89973-1G 89973-500MG
77630-854EA 1468 USD
77630-854 77636-244 77640-170 77648-150
2-Bromo-1-(cyclopropylmethoxy)-3-methoxybenzene ≥97%
2-Bromo-1-(cyclopropylmethoxy)-3-methoxybenzene

Precaución: For Research Use Only

Formula: 257.124 g/mol
MW: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD29095067
CAS Number: 1369905-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR