4-(Cyclopropylmethoxy)-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Cyclopropylmethoxy)benzoic acid

89907-5G 89907-25G 89907-1G 89907-500MG
77635-350EA 345 USD
77635-350 77657-042 77644-178 77652-176
4-(Cyclopropylmethoxy)-benzoic acid ≥97%
4-(Cyclopropylmethoxy)-benzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11156580
CAS Number: 355391-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR