2-(Benzyloxy)-3,5-dichloropyridine ≥95%

Supplier: AOBChem USA

Warning

89775-5G 89775-1G 89775-25G 89775-500MG
77633-826EA 912 USD
77633-826 77640-168 77657-040 77652-174
2-(Benzyloxy)-3,5-dichloropyridine ≥95%
2-(Benzyloxy)-3,5-dichloropyridine

Precaución: For Research Use Only

Formula: 254.112 g/mol
MW: C₁₂H₉Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD29044256
CAS Number: 1881295-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR