1-(3-Bromo-5-chloro-2-fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

89666-5G 89666-1G 89666-500MG 89666-25G
77630-848EA 1441 USD
77630-848 77640-164 77653-120 77660-506
1-(3-Bromo-5-chloro-2-fluorophenyl)cyclopentanol ≥95%
1-(3-Bromo-5-chloro-2-fluorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 293.56 g/mol
MW: C₁₁H₁₁BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34817974
CAS Number: 2969365-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR