1-(2,4,6-Trifluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

89500-5G 89500-1G 89500-25G 89500-500MG
77630-840EA 1441 USD
77630-840 77640-158 77657-036 77652-164
1-(2,4,6-Trifluorophenyl)cyclobutanol ≥95%
1-(2,4,6-Trifluorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD35165591
CAS Number: 3002442-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR