1-(2,4,6-Trifluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89490-5G 89490-25G 89490-1G 89490-500MG
77630-838EA 1441 USD
77630-838 77657-034 77644-172 77652-162
1-(2,4,6-Trifluorophenyl)cyclohexanol ≥95%
1-(2,4,6-Trifluorophenyl)cyclohexanol

Precaución: For Research Use Only

Formula: C₁₂H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34817968
CAS Number: 2918960-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR