1-(3-Bromo-2,6-difluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89245-5G 89245-1G 89245-500MG
77630-834EA 1553 USD
77630-834 77640-154 77648-144
1-(3-Bromo-2,6-difluorophenyl)cyclohexanol ≥95%
1-(3-Bromo-2,6-difluorophenyl)cyclohexanol

Precaución: For Research Use Only

Formula: C₁₂H₁₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34771857
CAS Number: 1699193-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR