1-(4-Methoxy-3,5-dimethylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89170-5G 89170-1G 89170-25G 89170-500MG
77630-832EA 1441 USD
77630-832 77640-150 77657-030 77652-156
1-(4-Methoxy-3,5-dimethylphenyl)cyclohexanol ≥95%
1-(4-Methoxy-3,5-dimethylphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 234.334 g/mol
MW: C₁₅H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34771848
CAS Number: 122004-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR