1-(2,3-Dimethylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89164-5G 89164-1G 89164-25G 89164-500MG
77630-830EA 1441 USD
77630-830 77640-146 77657-028 77648-142
1-(2,3-Dimethylphenyl)cyclohexanol ≥95%
1-(2,3-Dimethylphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 204.308 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD16794548
CAS Number: 854714-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR