1-(2,4-Difluorophenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89160-5G 89160-1G 89160-25G 89160-500MG
77630-828EA 1441 USD
77630-828 77640-144 77660-498 77652-152
1-(2,4-Difluorophenyl)cyclohexanol ≥97%
1-(2,4-Difluorophenyl)cyclohexanol

Precaución: For Research Use Only

Formula: C₁₂H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD17223577
CAS Number: 1341197-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR