1-(3-Fluoro-5-methoxyphenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89154-5G 89154-25G 89154-1G 89154-500MG
77630-826EA 1441 USD
77630-826 77657-026 77644-170 77652-150
1-(3-Fluoro-5-methoxyphenyl)cyclohexanol ≥97%
1-(3-Fluoro-5-methoxyphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 224.271 g/mol
MW: C₁₃H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34771844
CAS Number: 2003045-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR