1-(4-Phenoxyphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89119-5G 89119-25G 89119-1G 89119-500MG
77630-824EA 1441 USD
77630-824 77657-024 77644-168 77648-140
1-(4-Phenoxyphenyl)cyclohexanol ≥95%
1-(4-Phenoxyphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 268.35 g/mol
MW: C₁₈H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34771837
CAS Number: 2410095-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR