Ethyl 2-(6-bromo-2-fluoro-3-iodophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

89088-5G 89088-1G 89088-500MG
77660-496EA 2688 USD
77660-496 77644-166 77648-138
Ethyl 2-(6-bromo-2-fluoro-3-iodophenyl)-2-oxoacetate ≥95%
Ethyl 2-(6-bromo-2-fluoro-3-iodophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 400.968 g/mol
MW: C₁₀H₇BrFIO₃
Storage Temperature: Ambient
MDL Number: MFCD34771822
CAS Number: 2918887-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR