2-Bromo-4-chloro-3-isopropoxy-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

88694-5G 88694-1G 88694-25G 88694-500MG
77630-820EA 1441 USD
77630-820 77640-140 77657-018 77648-136
2-Bromo-4-chloro-3-isopropoxy-1-methylbenzene ≥95%
2-Bromo-4-chloro-3-isopropoxy-1-methylbenzene

Precaución: For Research Use Only

Formula: 263.559 g/mol
MW: C₁₀H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD34771643
CAS Number: 2901098-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR