1-(2-Isopropylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

88559-5G 88559-1G 88559-25G 88559-500MG
77630-816EA 1441 USD
77630-816 77640-138 77657-014 77652-146
1-(2-Isopropylphenyl)cyclobutanol ≥95%
1-(2-Isopropylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34771581
CAS Number: 1874917-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR