1-(2-Ethylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

88553-5G 88553-25G 88553-1G 88553-500MG
77630-814EA 1441 USD
77630-814 77657-012 77644-162 77652-144
1-(2-Ethylphenyl)cyclopentanol ≥95%
1-(2-Ethylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 190.281 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34771577
CAS Number: 1508537-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR