1-(2-Ethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

88552-5G 88552-1G 88552-25G 88552-500MG
77630-812EA 1441 USD
77630-812 77640-136 77660-488 77648-132
1-(2-Ethylphenyl)cyclobutanol ≥95%
1-(2-Ethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 176.255 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD30163075
CAS Number: 1862912-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR