1-(2-Ethylphenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

88551-5G 88551-25G 88551-1G 88551-500MG
77630-810EA 1441 USD
77630-810 77657-010 77644-160 77652-142
1-(2-Ethylphenyl)-2-fluoroethanone ≥95%
1-(2-Ethylphenyl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD34771576
CAS Number: 2918928-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR