1-Bromo-3,5-dichloro-4-methoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

88442-5G 88442-1G 88442-25G 88442-500MG
77630-808EA 1441 USD
77630-808 77640-134 77660-484 77652-140
1-Bromo-3,5-dichloro-4-methoxy-2-methylbenzene ≥95%
1-Bromo-3,5-dichloro-4-methoxy-2-methylbenzene

Precaución: For Research Use Only

Formula: C₈H₇BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34771512
CAS Number: 2901109-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR