2-Iodo-3-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

88380-5G 88380-25G
77635-348EA 175 USD
77635-348 77660-482
2-Iodo-3-methylbenzamide ≥95%
2-Iodo-3-methylbenzamide

Precaución: For Research Use Only

Formula: C₈H₈INO
MW: 261.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753716
CAS Number: 887234-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR