1-Bromo-2-chloro-3-methyl-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

87653-5G 87653-25G 87653-1G
77635-346EA 186 USD
77635-346 77660-476 77644-158
1-Bromo-2-chloro-3-methyl-5-nitrobenzene ≥97%
1-Bromo-2-chloro-3-methyl-5-nitrobenzene

Precaución: For Research Use Only

Formula: 250.477 g/mol
MW: C₇H₅BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD11846020
CAS Number: 116277-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR